Tisk…

1-Heptanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 1-heptanesulphonate monohydrate

51835-10G 51835-250G 51835-50G
SIAL51835-10GEA 2190 CZK
SIAL51835-10G SIAL51835-250G SIAL51835-50G
1-Heptanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®
1-Heptanesulphonic acid sodium salt monohydrate
Vzorec: C₇H₁₅NaO₃S·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS číslo: 207300-90-1
EINECS: 245-210-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR