Tisk…

1-Heptanesulphonic acid sodium salt monohydrate, Supelco®

Dodavatel: Merck

Synonyms: Sodium 1-heptanesulphonate monohydrate

M-Clarity™ quality level = MQ100

51832-10G-F 51832-50G-F
51832-10G-FEA 12770 CZK
51832-10G-F 51832-50G-F
1-Heptanesulphonic acid sodium salt monohydrate, Supelco®
1-Heptanesulphonic acid sodium salt monohydrate
Vzorec: C₇H₁₅NaO₃S·1H₂O
MW: 220.27
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS číslo: 207300-90-1
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc61ea40aea34a4ecc116221e6b8f64cb4dd245&vid=6423e1e17128215a02c27a62b05b8e4454209037&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR