EDTA čtyřsodná sůl hydrát, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tetrasodium ethylenediaminetetraacetate hydrate, Kestranal® 4S hydrate, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate, Edetic acid tetrasodium salt hydrate, Ethylenediaminetetraacetic acid tetrasodium salt hydrate
M-Clarity™ quality level = MQ200
Vzorec:
C₁₀H₁₂N₂O₈Na₄·xH₂O MW: 380,17 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150025 CAS číslo: 13235-36-4 EINECS: 200-573-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...