Ethylendiamintetraacetát tetrasodný dihydrát, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tetrasodium ethylenediaminetetraacetate dihydrate, Edetic acid tetrasodium salt dihydrate, Kestranal® 4S 2 H2O, Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetraacetic acid sodium hydrate, Tetrasodium Ethylenediaminetetraacetate Dihydrate, Kestranal® 4S dihydrate
M-Clarity™ quality level = MQ200
ED4SS-100G
ED4SS-1KG
ED4SS-2.5KG
ED4SS-500G
SIALED4SS-100GEA
2310
CZK
SIALED4SS-100G
SIALED4SS-1KG
SIALED4SS-2.5KG
SIALED4SS-500G
Ethylendiamintetraacetát tetrasodný dihydrát, Sigma-Aldrich®
Ethylendiamintetraacetát tetrasodný dihydrát
Vzorec:
C₁₀H₁₂N₂Na₄O₈·2H₂O MW: 416,2 g/mol Bod tání: > 300 °C Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150026 CAS číslo: 10378-23-1 EINECS: 200-573-9 Merck Index: 13,03544 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...