Tisk…
EDTA dvojsodná sůl dihydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
M-Clarity™ quality level = MQ200
E1644-100G E1644-1KG E1644-250G
SIALE1644-100GEA 2980 CZK
SIALE1644-100G SIALE1644-1KG SIALE1644-250G
EDTA dvojsodná sůl dihydrát, Sigma-Aldrich®
EDTA dvojsodná sůl dihydrát
Vzorec: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24
Bod tání: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS číslo: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM