Tisk…
Citronan sodný dihydrát ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
A12274.0C A12274.0E A12274.36
A12274.0CEA 9130 CZK
A12274.0C A12274.0E A12274.36
Citronan sodný dihydrát ≥99%
Citran trojdraselný dihydrát
Purity: 99% CAS No.: 6132-04-3 Molecular Formula: Na3C6H5O7.2H2O Formula Weight: 294.10 (258.07anhy) Odour: Odourless Melting Point: 150° -2H2O Beilstein Reference: 6104939 Merck Reference: 14,8602 MDL No.: MFCD00150031
Vzorec: Na₃C₆H₅O₇·2H₂O
MW: 294.1
Bod tání: 150 °C (-2H₂O)
Hustota: 1.76
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS číslo: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM