Nikl-acetát tetrahydrát ≥99.999% (metals basis), Puratronic®
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate
Purity: 99.999%
CAS No.: 6018-89-9
Molecular Formula: C4H6NiO4.4H2O
Formula Weight: 248.84 (176.79anhy)
Physical Form: Crystalline Aggregates
Density: 1.744
Appearance: Green
Odour: Light
Melting Point: dec.
Beilstein Reference: 3730764
Merck Reference: 14,6499
UN No.: 3077
MDL No.: MFCD00066973
Vzorec:
C₄H₆NiO₄·4H₂O MW: 248,84 g/mol Bod varu: 16 °C (1013 hPa) Bod tání: Dec. Hustota: 1,744 Storage Temperature: Ambient |
MDL Number:
MFCD00066973 CAS číslo: 6018-89-9 EINECS: 206-761-7 UN: 3077 ADR: 9,III Merck Index: 14,6499 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...