Tisk…

Octan měďnatý monohydrát ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

A16203.30 A16203.0B A16203.0I
A16203.30EA 1340 CZK
A16203.30 A16203.0B A16203.0I
Octan měďnatý monohydrát ≥98%
Octan měďnatý monohydrát

Purity: 98+% CAS No.: 6046-93-1 Molecular Formula: C4H6CuO4.H2O Formula Weight: 199.65 (181.64anhy) Physical Form: Powder Density: 1.882 Appearance: Blue-green Odour: Odourless pH: 5.2-5.5 Melting Point: 115° Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00149570

Vzorec: (H₃CCOO)₂Cu·H₂O
MW: 199.65
Bod tání: 115
Hustota: 1.882
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS číslo: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9 III
Merck Index: 13,02651

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR