Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite ≥95%
Dodavatel: Merck
766305-1G
766305-5G
SIAL766305-1GEA
4980
CZK
SIAL766305-1G
SIAL766305-5G
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite ≥95%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite
Useful phosphorylating reagent used in oligonucleotide synthesis for adding a terminal phosphate group to the 3′ or 5′ hydroxyl.
Vzorec:
C₁₂H₂₂N₃O₂P |
MDL Number:
MFCD00797597 CAS číslo: 102690-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...