Tisk…

Fenylhydraziniumchlorid ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

A14645.22 A14645.36
A14645.22EA 2150 CZK
A14645.22 A14645.36
Fenylhydraziniumchlorid ≥99%
Fenylhydraziniumchlorid

Purity: 99% CAS No.: 59-88-1 Molecular Formula: C6H8N2.HCl Formula Weight: 144.61 Melting Point: ca 252° dec. Beilstein Reference: 3594958 Merck Reference: 14,7293 UN No.: 2811 MDL No.: MFCD00012924

Vzorec: C₆H₉ClN₂
MW: 144,6 g/mol
Bod tání: ∼ 252 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS číslo: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,7293
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR