Tisk…

Hydrazin dihydrochlorid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride

M-Clarity™ quality level = MQ100

216208-100G 216208-500G
SIAL216208-100GEA 1870 CZK
SIAL216208-100G SIAL216208-500G
Hydrazin dihydrochlorid, Sigma-Aldrich®
Hydrazin dihydrochlorid
Vzorec: H₆Cl₂N₂
MW: 104,97 g/mol
Bod tání: 189 °C
Hustota: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00064543
CAS číslo: 5341-61-7
EINECS: 226-283-2
UN: 2928
ADR: 6.1,II
Merck Index: 13,04789

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR