Tisk…

Hydrazin dihydrochlorid 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride

411761000 411765000 411760025
ACRO411761000EA 1730 CZK
ACRO411761000 ACRO411765000 ACRO411760025
Hydrazin dihydrochlorid 99%
Hydrazin dihydrochlorid
Vzorec: H₆Cl₂N₂
MW: 104,97 g/mol
Bod tání: 198 °C
Hustota: 1,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064543
CAS číslo: 5341-61-7
EINECS: 226-283-2
UN: 3288
ADR: 6.1,III
Merck Index: 13,04789

Objednat

Výsledky specifikačního testu

Appearance White Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR