Hydraziniumsulfát ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10792.30
A10792.A1
A10792.30EA
1140
CZK
A10792.30
A10792.A1
Hydraziniumsulfát ≥99%
Hydraziniumsulfát
Purity: 99+%
CAS No.: 10034-93-2
Molecular Formula: H2NNH2.H2SO4
Formula Weight: 130.12
Odour: Odourless
pH: 1.3
Melting Point: 254°
Merck Reference: 14,4772
UN No.: 3288
MDL No.: MFCD00044873
Vzorec:
NH₂NH₂·H₂SO₄ Bod tání: 254 °C Hustota: 1,370 Storage Temperature: Ambient |
MDL Number:
MFCD00044873 CAS číslo: 10034-93-2 EINECS: 233-110-4 UN: 3288 ADR: 6.1,III Merck Index: 14,4772 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...