Tisk…

Hydrazin-Hydrat 98+%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Hydrazinium hydroxide, Hydrazine monohydrate

A14005.36 A14005.0E A14005.18 A14005.30
A14005.36EA 1740 CZK
A14005.36 A14005.0E A14005.18 A14005.30
Hydrazin-Hydrat 98+%
Hydrazin-Hydrat

Purity: 98+% CAS No.: 7803-57-8 Molecular Formula: H4N2.H2O Formula Weight: 50.06 (32.05anhy) Flash Point: 75°(167°F) Refractive Index: 1.4310 Odour: Amine-like Melting Point: -52° Boiling Point: 120-121° Merck Reference: 14,4771 UN No.: 2030 MDL No.: MFCD00149931

Vzorec: H₆N₂O
MW: 50,06 g/mol
Bod varu: 120…121 °C
Bod tání: –52 °C
Hustota: 1,027
Bod vzplanutí: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149931
CAS číslo: 7803-57-8
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 14,4771

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR