Tisk…

Hydrazin-Hydrat 99+%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

16651.22 16651.36 16651.A3 16651.57
16651.22EA 1740 CZK
16651.22 16651.36 16651.A3 16651.57
Hydrazin-Hydrat 99+%
Hydrazin-Hydrat

Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states.


  • 64% H₂NNH₂ in H₂O


It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

Vzorec: NH₂NH₂·xH₂O
Bod varu: 120…121 °C
Bod tání: –52 °C
Hustota: 1,027
Bod vzplanutí: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149931
CAS číslo: 10217-52-4
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 14,4771

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR