Tisk…

1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Dodavatel: Thermo Scientific
Varování

Synonyms: Acetylene-PEG4-amine, 3,6,9,12-Tetraoxapentadec-14-yn-1-amine, Propargyl-PEG4-amine

J64527.MC J64527.ME
J64527.MCEA 12430 CZK
J64527.MC J64527.ME
1-Amino-3,6,9,12-tetraoxapentadec-14-yne
1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Acetylene containing reagent with PEG spacer and carboxyl reactivity.

Vzorec: C₁₁H₂₁NO₄
MW: 231,29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22380736
CAS číslo: 1013921-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR