Tisk…

Guanidinium nitrate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Guanidine nitrate

L09643.30 L09643.0B
L09643.30EA 1240 CZK
L09643.30 L09643.0B
Guanidinium nitrate ≥98%
Guanidinium nitrate

Purity: 98% CAS No.: 506-93-4 Molecular Formula: CH6N4O3 Formula Weight: 122.08 Odour: Odourless pH: 4.9 Melting Point: 213-216° Beilstein Reference: 3596600 Merck Reference: 14,4562 UN No.: 1467 MDL No.: MFCD00013028

Vzorec: CH₆N₄O₃
MW: 122,08 g/mol
Bod tání: 213…216 °C
Hustota: 1,44
Storage Temperature: Ambient
MDL Number: MFCD00013028
CAS číslo: 506-93-4
EINECS: 208-060-1
UN: 1467
ADR: 5.1,III
Merck Index: 14,4562

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR