Tisk…
Lysates, Jurkat (human)
Dodavatel: Enzo Life Sciences
ENZOADILYCJK100FEA 4490 CZK
ALEXADILYCJK100F ENZOADILYCJK100F
Lysates, Jurkat (human)
Buňky
The Jurkat T lymphocyte cell line was established from the peripheral blood of a 14-year old boy with T cell leukaemia.
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer, pH 6,8, heated for 10 minutes at 70 °C and containing Tris-HCl (40 mM), SDS (1%), DTT (50 mM), glycerol (7,5%), and bromophenol blue (0,003%).