Tisk…

Nový fuchsin C.I. 42520

Dodavatel: MP Biomedicals
Varování

Synonyms: Basic Violet 2, New Fuchsin, 4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

0215582380 0215582325
ICNA0215582380EA 2820 CZK
ICNA0215582380 ICNA0215582325
Nový fuchsin C.I. 42520
Nový fuchsin

Soluble in water and ethanol.

Vzorec: C₂₂H₂₄ClN₃
MW: 365,91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012567
CAS číslo: 3248-91-7
EINECS: 221-831-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR