4-(Methylamino)phenol sulphate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Metol, Bis(4-hydroxy-N-methylanilinium)sulfát
M-Clarity™ quality level = MQ200
69750-100G
69750-500G
SIAL69750-100GEA
1480
CZK
SIAL69750-100G
SIAL69750-500G
4-(Methylamino)phenol sulphate, Sigma-Aldrich®
Bis(4-hydroxy-N-methylanilinium)sulfát
Vzorec:
C₁₄H₂₀N₂O₆S MW: 344,39 g/mol Bod varu: 227 °C (1013 hPa) Bod tání: 86 °C Hustota: 1,434 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013140 CAS číslo: 55-55-0 EINECS: 200-237-1 UN: 2811 ADR: 6.1,III Merck Index: 13,06046 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...