Tisk…
N,N-Dimethyl-1,4-fenylendiamoniumdichlorid, Millipore®
Dodavatel: Merck
Nebezpečí
Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl
M-Clarity™ quality level = MQ200
07770-100G 07770-25G
SIAL07770-100GEA 0 CZK
SIAL07770-100G SIAL07770-25G
N,N-Dimethyl-1,4-fenylendiamoniumdichlorid, Millipore®
N,N-Dimethyl-1,4-fenylendiamoniumdichlorid
Vzorec: (CH₃)₂NC₆H₄NH₂·2HCl
Bod tání: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00012991
CAS číslo: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280