Tisk…
Spermidine trihydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride
M-Clarity™ quality level = MQ200
S2501-1G S2501-25G S2501-5G
SIALS2501-1GEA 1250 CZK
SIALS2501-1G SIALS2501-25G SIALS2501-5G
Spermidine trihydrochloride, Sigma-Aldrich®
Spermidine trihydrochloride
Vzorec: C₇H₂₂Cl₃N₃
MW: 254,63 g/mol
Bod tání: 256…258 °C
Storage Temperature: Freezer
MDL Number: MFCD00012918
CAS číslo: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893