Tisk…

Diethylamine hydrochloride 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diethylammonium chloride

A13637.0B A13637.0I A13637.30
A13637.0BEA 3110 CZK
A13637.0B A13637.0I A13637.30
Diethylamine hydrochloride 99%
Diethylamine hydrochloride

Purity: 99% CAS No.: 660-68-4 Molecular Formula: (CH3CH2)2NH.HCl Formula Weight: 109.60 Melting Point: 227-230° Boiling Point: 320-330° Merck Reference: 14,3111 MDL No.: MFCD00012499

Vzorec: C₄H₁₂ClN
MW: 109.6
Bod varu: 320…330 °C
Bod tání: 227…230 °C
Hustota: 1.0475 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012499
CAS číslo: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR