Tisk…
L(+)-cystein hydrochlorid monohydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
M-Clarity™ quality level = MQ100
C121800-100G C121800-5G
SIALC121800-100GEA 2660 CZK
SIALC121800-100G SIALC121800-5G
L(+)-cystein hydrochlorid monohydrát, Sigma-Aldrich®
L(+)-cystein hydrochlorid monohydrát
Vzorec: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64
Bod tání: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS číslo: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM