Tisk…
Paladium 5% on barium sulphate, powder hydrogenation catalyst, reduced
Dodavatel: Thermo Fisher Scientific
11722.14 11722.22 11722.88
11722.14EA 14210 CZK
11722.14 11722.22 11722.88
Paladium 5% on barium sulphate, powder hydrogenation catalyst, reduced
Palladium
Appearance: Grey-black MDL No.: MFCD03613605
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6