Tisk…

Tetraethylammonium hydroxide 40% vodný roztok, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Tetrylammonium hydroxide

86633-100ML 86633-500ML
SIAL86633-100MLEA 0 CZK
SIAL86633-100ML SIAL86633-500ML
Tetraethylammonium hydroxide 40% vodný roztok, Sigma-Aldrich®
Tetraethylammonium hydroxide
Vzorec: C₈H₂₁NO
MW: 147,26 g/mol
MDL Number: MFCD00009024
CAS číslo: 77-98-5
EINECS: 201-073-3
UN: 3267
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR