Tisk…
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
M-Clarity™ quality level = MQ200
178780-250ML 178780-50ML 178780-1L 178780-2.5L
SIAL178780-250MLEA 5840 CZK
SIAL178780-250ML SIAL178780-50ML SIAL178780-1L SIAL178780-2.5L
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Tetrabutylammonium hydroxide
Vzorec: C₁₆H₃₇NO
MW: 259.48
Bod tání: 29.85
Hustota: 0.968
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS číslo: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II