Tisk…
thiokyanatan amonný solution_standardised 0.1 N aqueous solution
Dodavatel: Thermo Fisher Scientific
Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
35580.K2 35580.K7
35580.K2EA 1590 CZK
35580.K2 35580.K7
thiokyanatan amonný solution_standardised 0.1 N aqueous solution
thiokyanatan amonný
Molecular Formula: NH4SCN Formula Weight: 76.12 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00011428
Vzorec: NH₄SCN
MW: 76.12
Bod tání: 150
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS číslo: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564