Tisk…
Diisobutylaluminium hydride 1 mol/l in toluene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
M-Clarity™ quality level = MQ200
215007-100ML 215007-18L 215007-800ML
SIAL215007-100MLEA 2360 CZK
SIAL215007-100ML SIAL215007-18L SIAL215007-800ML
Diisobutylaluminium hydride 1 mol/l in toluene, Sigma-Aldrich®
Diisobutylaluminium hydride
Vzorec: C₈H₁₉Al
MW: 142.22
Bod tání: -80
Hustota: 0.798
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS číslo: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3 I