Tisk…

p-Toluenesulphonic acid 12% in acetic acid, pure

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Toluene-4-sulphonic acid

421220010 421222500
ACRO421220010EA 7250 CZK
ACRO421220010 ACRO421222500
p-Toluenesulphonic acid 12% in acetic acid, pure
p-Toluenesulphonic acid
Vzorec: C₇H₈O₃S
MW: 172,2 g/mol
Bod vzplanutí: 41 °C
CAS číslo: 104-15-4
EINECS: 203-180-0
UN: 2920

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow to pink Solution
Titration with NaOH 11 to 13 %
Melting point 84 to 88 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR