ethylamin ≥70%, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Monoethylamine, Aminoethane, Ethanamine
M-Clarity™ quality level = MQ200
471208-100ML
471208-500ML
SIAL471208-100MLEA
1030
CZK
SIAL471208-100ML
SIAL471208-500ML
ethylamin ≥70%, Sigma-Aldrich®
ethylamin
Vzorec:
C₂H₇N MW: 45,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008160 CAS číslo: 75-04-7 EINECS: 200-834-7 UN: 2270 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...