Tisk…
EDTA disodium salt 0.01 mol/l (0.02 N) aqueous solution
Dodavatel: REAGECON
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
ED20011 ED20015
REGEED20011EA 872 CZK
REGEED20011 REGEED20015
EDTA disodium salt 0.01 mol/l (0.02 N) aqueous solution
EDTA disodium salt
Vzorec: C₁₀H₁₄N₂Na₂O₈
MW: 336.21
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS číslo: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556