(±)-3-Carene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W382108-1KG
W382108-9KG
W382108-SAMPLE
SIALW382108-1KGEA
0
CZK
SIALW382108-1KG
SIALW382108-9KG
SIALW382108-SAMPLE
(±)-3-Carene, Sigma-Aldrich®
(±)-3-Carene
Vzorec:
C₁₀H₁₆ Bod varu: 167…170 °C (1013 hPa) Hustota: 0,86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001315 CAS číslo: 13466-78-9 EINECS: 236-719-3 UN: 2319 ADR: 3,III Merck Index: 12,01885 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...