(±)-2-Phenyl-1-propanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Phenylpropan-1-ol
M-Clarity™ quality level = MQ300
W273201-1KG
W273201-5KG
W273201-SAMPLE
SIALW273201-1KGEA
25480
CZK
SIALW273201-1KG
SIALW273201-5KG
SIALW273201-SAMPLE
(±)-2-Phenyl-1-propanol, Sigma-Aldrich®
(±)-2-Phenyl-1-propanol
Vzorec:
C₆H₅CH(CH₃)CH₂OH MW: 136,19 g/mol Bod varu: 110 °C (1013 hPa) Bod tání: –37 °C Hustota: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004736 CAS číslo: 1123-85-9 EINECS: 214-379-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...