(±)-2-Phenyl-1-propanol ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Phenylpropan-1-ol
B23473.18
B23473.30
B23473.18EA
1590
CZK
B23473.18
B23473.30
(±)-2-Phenyl-1-propanol ≥97%
(±)-2-Phenyl-1-propanol
Purity: 97%
CAS No.: 1123-85-9
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 108°(226°F)
Refractive Index: 1.5262
Melting Point: -37°
Boiling Point: 220°
Beilstein Reference: 1906760
MDL No.: MFCD00004736
Vzorec:
C₆H₅CH(CH₃)CH₂OH MW: 136,19 g/mol Bod varu: 220 °C Bod tání: –37 °C Hustota: 0,975 Bod vzplanutí: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004736 CAS číslo: 1123-85-9 EINECS: 214-379-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...