(±)-1-Fenylethanol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol
M-Clarity™ quality level = MQ300
W268518-10KG-K
W268518-1KG-K
W268518-25KG-K
SIALW268518-10KG-KEA
29000
CZK
SIALW268518-10KG-K
SIALW268518-1KG-K
SIALW268518-25KG-K
(±)-1-Fenylethanol, Sigma-Aldrich®
(±)-1-Fenylethanol
Vzorec:
C₆H₅CH(OH)CH₃ MW: 122.17 Bod varu: 401 Bod tání: 20 Hustota: 1.015 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004508 CAS číslo: 98-85-1 EINECS: 202-707-1 UN: 2937 ADR: 6.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...