(±)-3-Butyne-2-ol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A11881.09
A11881.06
A11881.14
A11881.22
A11881.09EA
1890
CZK
A11881.09
A11881.06
A11881.14
A11881.22
(±)-3-Butyne-2-ol ≥98%
(±)-3-Butyne-2-ol
Purity: 98%
CAS No.: 2028-63-9
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 28°(82°F)
Refractive Index: 1.4260
Boiling Point: 108-109°
Beilstein Reference: 635722
UN No.: 2929
MDL No.: MFCD00004541
Vzorec:
C₄H₆O Bod varu: 108…109 °C Bod tání: –12 °C Hustota: 0,894 Bod vzplanutí: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004541 CAS číslo: 2028-63-9 EINECS: 217-978-1 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...