(±)-4-Penten-2-ol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B20208.03
B20208.06
B20208.14
B20208.03EA
906
CZK
B20208.03
B20208.06
B20208.14
(±)-4-Penten-2-ol ≥98%
(±)-4-Penten-2-ol
Purity: 98%
CAS No.: 625-31-0
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 30°(86°F)
Refractive Index: 1.4240
Boiling Point: 115-116°
UN No.: 1987
MDL No.: MFCD00004556
Vzorec:
C₅H₁₀O Bod varu: 115…116 °C Hustota: 0,837 Bod vzplanutí: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004556 CAS číslo: 625-31-0 EINECS: 210-887-8 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...