Tisk…

(±)-3-Buten-2-ol ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L14787.06 L14787.14
L14787.06EA 1040 CZK
L14787.06 L14787.14
(±)-3-Buten-2-ol ≥97%
(±)-3-Buten-2-ol

Purity: 97% CAS No.: 598-32-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: 20°(68°F) Refractive Index: 1.4140 Melting Point: -100° Boiling Point: 96-98° Beilstein Reference: 1361410 UN No.: 1986 MDL No.: MFCD00004543

Vzorec: C₄H₈O
Bod varu: 96…98 °C
Bod tání: < –100 °C
Hustota: 0,838
Bod vzplanutí: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004543
CAS číslo: 598-32-3
EINECS: 209-929-8
UN: 1986
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR