(±)-Phenylsuccinic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
P35200-100G
P35200-25G
SIALP35200-100GEA
2700
CZK
SIALP35200-100G
SIALP35200-25G
(±)-Phenylsuccinic acid, Sigma-Aldrich®
(±)-Phenylsuccinic acid
Vzorec:
C₁₀H₁₀O₄ Bod varu: 368 °C (1013 hPa) Bod tání: 167…169 °C |
MDL Number:
MFCD00004256 CAS číslo: 635-51-8 EINECS: 211-238-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...