2-Phenylpropionic acid ≥97%, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
(±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
M-Clarity™ quality level = MQ100
P31701-10G
P31701-5G
SIALP31701-10GEA
2190
CZK
SIALP31701-10G
SIALP31701-5G
2-Phenylpropionic acid ≥97%, Sigma-Aldrich®
2-Phenylpropionic acid
Vzorec:
C₉H₁₀O₂ MW: 150,18 g/mol Bod varu: 260 °C (1013 hPa) Bod tání: –5 °C Hustota: 1,1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002650 CAS číslo: 492-37-5 EINECS: 207-752-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...