DL-Tryptofan, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
H-DL-Trp-OH, (+/-)-2-Amino-3-(3-indolyl)propionic acid, 2-Amino-3-(1H-indol-3-yl)propanoic acid
M-Clarity™ quality level = MQ200
T3300-100G
T3300-25G
T3300-5G
SIALT3300-100GEA
25730
CZK
SIALT3300-100G
SIALT3300-25G
SIALT3300-5G
DL-Tryptofan, Sigma-Aldrich®
DL-Tryptofan
Vzorec:
C₁₁H₁₂N₂O₂ MW: 204.23 Bod tání: 289…290 °C Hustota: 1.349 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064339 CAS číslo: 54-12-6 EINECS: 200-194-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...