Tisk…

DL-Histidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Amino-3-(1H-imidazol-4-yl)propanoic acid, H-DL-His-OH

M-Clarity™ quality level = MQ200

H7750-100G H7750-25G
SIALH7750-100GEA 29370 CZK
SIALH7750-100G SIALH7750-25G
DL-Histidine, Sigma-Aldrich®
DL-Histidine
Vzorec: C₆H₉N₃O₂
MW: 155,16 g/mol
Bod tání: 273 °C
MDL Number: MFCD00005208
CAS číslo: 4998-57-6
EINECS: 206-513-8
Merck Index: 12,04758

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR