Tisk…

(±)-2-methyl-2,4-pentandiol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol

L03187.AE L03187.AP L03187.0F
L03187.AEEA 803 CZK
L03187.AE L03187.AP L03187.0F
(±)-2-methyl-2,4-pentandiol ≥98%
(±)-2-methyl-2,4-pentandiol

Purity: 98% CAS No.: 107-41-5 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 93°(199°F) Refractive Index: 1.4270 Odour: Weak Melting Point: -40° Boiling Point: 197° Beilstein Reference: 1098298 Merck Reference: 14,4710 MDL No.: MFCD00004547

Vzorec: C₆H₁₄O₂
MW: 118.18
Bod varu: 197 °C
Bod tání: –40 °C
Hustota: 0.922
Bod vzplanutí: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS číslo: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR