(±)-3-Methyl-1-pentanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methylpentan-1-ol, (±)-3-Methylpentan-1-ol, 3-Methyl-1-pentanol
M-Clarity™ quality level = MQ300
W376205-100G-K
W376205-1KG-K
SIALW376205-100G-KEA
0
CZK
SIALW376205-100G-K
SIALW376205-1KG-K
(±)-3-Methyl-1-pentanol, Sigma-Aldrich®
(±)-3-Methyl-1-pentanol
Vzorec:
C₆H₁₄O Bod varu: 152…153 °C (1013 hPa) Bod tání: –70 °C Hustota: 0,823 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002937 CAS číslo: 589-35-5 EINECS: 209-644-9 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...