Tisk…

(±)-2-Methyl-1-pentanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methylamyl alcohol, 2-Methyl-1-pentanol, 2-Propylpropanol

M-Clarity™ quality level = MQ100

214019-50ML 214019-250ML
SIAL214019-50MLEA 2470 CZK
SIAL214019-50ML SIAL214019-250ML
(±)-2-Methyl-1-pentanol, Sigma-Aldrich®
(±)-2-Methyl-1-pentanol
Vzorec: C₆H₁₄O
MW: 102.18
Bod varu: 148
Bod tání: 0
Hustota: 0.82
Storage Temperature: Ambient
MDL Number: MFCD00004745
CAS číslo: 105-30-6
EINECS: 203-285-1
UN: 1987
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR