Tisk…

(±)-2-Etyl-1-hexanol ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Ethylhexan-1-ol

A17104.0F A17104.AP
A17104.0FEA 2110 CZK
A17104.0F A17104.AP
(±)-2-Etyl-1-hexanol ≥99%
(±)-2-Etyl-1-hexanol

Purity: 99% CAS No.: 104-76-7 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 77°(171°F) Refractive Index: 1.4310 Odour: Mild Melting Point: -76° Boiling Point: 183-186° Beilstein Reference: 1719280 Merck Reference: 14,3808 MDL No.: MFCD00004746

Vzorec: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MW: 130,23 g/mol
Bod varu: 183…186 °C
Bod tání: –76 °C
Hustota: 0,833
Bod vzplanutí: 77 °C (171 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS číslo: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 14,3808
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc81f9275fbd3a7f8fb0e453419eab1652a4fdec&vid=1f7059eb6b27d30ac2c7e6b83f6bc0dff6002fe7&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR