Tisk…

DL-Leucin 99+%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-Amino-4-methylpentanoic acid, DL-Leucine, (±)-Leucine

172130250 172132500
ACRO172130250EA 1330 CZK
ACRO172130250 ACRO172132500
DL-Leucin 99+%
DL-Leucin
Vzorec: C₆H₁₃NO₂
MW: 131.17
Bod varu: 226
Bod tání: 293…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00063087
CAS číslo: 328-39-2
EINECS: 206-328-2
Merck Index: 13,05470

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.1 %
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR