Tisk…

DL-Valin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-DL-Val-OH, (±)-Valine, 2-Amino-3-methylbutanoic acid

94640-100G 94640-25G 94640-500G
SIAL94640-100GEA 2280 CZK
SIAL94640-100G SIAL94640-25G SIAL94640-500G
DL-Valin, Sigma-Aldrich®
DL-Valin
Vzorec: C₅H₁₁NO₂
MW: 117,15 g/mol
Bod varu: 217 °C (1013 hPa)
Bod tání: 86 °C
Hustota: 1,352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS číslo: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR