Tisk…

(±)-2-Methylvaleric acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (±)-2-Methylpentanoic acid, 2-Methylpentanoic Acid, 2-Methylvaleric Acid

A13873.0E A13873.22 A13873.36
A13873.0EEA 19050 CZK
A13873.0E A13873.22 A13873.36
(±)-2-Methylvaleric acid ≥98%
(±)-2-Methylvaleric acid

Purity: 98+% CAS No.: 97-61-0 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 91°(195°F) Refractive Index: 1.4140 Odour: Unpleasant Melting Point: -85° Boiling Point: 196-197° Beilstein Reference: 1720655 UN No.: 3265 MDL No.: MFCD00002671

Vzorec: C₆H₁₂O₂
MW: 116,16 g/mol
Bod varu: 196…197 °C
Bod tání: –85 °C
Hustota: 0,930
Bod vzplanutí: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002671
CAS číslo: 97-61-0
EINECS: 202-594-9
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR